2-Bromo-2'-(trifluoromethyl)acetophenone

Supplier: APOLLO SCIENTIFIC
Danger

PC5799-10G PC5799-1G PC5799-5G
77417-092EA 309.72 USD
77417-092 77417-094 77417-096
2-Bromo-2'-(trifluoromethyl)acetophenone
2-Bromo-2'-(trifluoromethyl)acetophenone
Formula: C₉H₆BrF₃O
Boiling Pt: 52 °C/0.03 mmHg
Melting Pt: 24…32 °C
Storage Temperature: Ambient
MDL Number: MFCD03094304
CAS Number: 54109-16-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR