2-Chloro-1,1,1,6,6,6-hexafluorohexa-2,4-diene

Supplier: APOLLO SCIENTIFIC
Warning

PC5778-1G PC5778-5G
77417-054EA 149.76 USD
77417-054 77417-056
2-Chloro-1,1,1,6,6,6-hexafluorohexa-2,4-diene
2-Chloro-1,1,1,6,6,6-hexafluorohexa-2,4-diene
Formula: C₆H₃ClF₆
Storage Temperature: Ambient
MDL Number: MFCD08461616
CAS Number: 885276-10-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR