Trifluoro(pyridine)boron 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC56215-1G PC56215-250MG PC56215-5G
77416-826EA 771.32 USD
77416-826 77416-828 77416-830
Trifluoro(pyridine)boron 97%
Trifluoro(pyridine)boron
Formula: C₅H₅BF₃N
Melting Pt: 39…41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00971931
CAS Number: 407-76-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR