2-Amino-4-[3-(trifluoromethyl)phenyl]-1,3-thiazole 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(3-(Trifluoromethyl)phenyl)thiazol-2-amine

PC5619-1G PC5619-500MG
77416-818EA 28.36 USD
77416-818 77416-820
2-Amino-4-[3-(trifluoromethyl)phenyl]-1,3-thiazole 97%
2-Amino-4-[3-(trifluoromethyl)phenyl]-1,3-thiazole
Formula: C₁₀H₇F₃N₂S
Storage Temperature: Ambient
MDL Number: MFCD03034765
CAS Number: 172848-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR