2-Chloro-5-(trifluoromethyl)pyrimidine 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC5409-250MG
PC5409-5G
PC5409-1G
77416-734EA
29.54
USD
77416-734
77416-736
77416-732
2-Chloro-5-(trifluoromethyl)pyrimidine 97%
2-Chloro-5-(trifluoromethyl)pyrimidine
Formula:
C₅H₂ClF₃N₂ MW: 182.53 g/mol Melting Pt: 47…52 °C Flash Pt: 71 °C Storage Temperature: Freezer |
MDL Number:
MFCD09753482 CAS Number: 69034-12-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...