(2R)-2-Amino-1,1,1-trifluoropropane 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (2R)-1,1,1-Trifluoropropan-2-amine

PC53181-1G PC53181-250MG
77416-654EA 511.44 USD
77416-654 77416-656
(2R)-2-Amino-1,1,1-trifluoropropane 99%
(2R)-2-Amino-1,1,1-trifluoropropane
Formula: C₃H₆F₃N
MW: 113.082 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16817687
CAS Number: 779303-24-1
UN: 2733
ADR: 3+8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR