2-(Trifluoromethyl)pyridine

Supplier: APOLLO SCIENTIFIC
Danger

PC5273-1G PC5273-25G PC5273-5G
77416-572EA 21.26 USD
77416-572 77416-574 77416-576
2-(Trifluoromethyl)pyridine
2-(Trifluoromethyl)pyridine
Formula: C₆H₄F₃N
Boiling Pt: 139…141 °C/754 mmHg
Density: 1.275 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02183562
CAS Number: 368-48-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR