2,5-Bis(trifluoromethyl)toluene 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 2-Methyl-1,4-bis(trifluoromethyl)benzene

PC52480-10G PC52480-1G PC52480-5G
77416-560EA 544.04 USD
77416-560 77416-562 77416-564
2,5-Bis(trifluoromethyl)toluene 98%
2,5-Bis(trifluoromethyl)toluene
Formula: C₉H₆F₆
MW: 228.134 g/mol
Storage Temperature: Ambient
CAS Number: 2385323-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR