Perfluorobutane-1-sulfonic anhydride 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,1,2,2,3,3,4,4,4-Nonafluorobutane-1-sulphonic anhydride, Nonafluorobutane-1-sulphonic anhydride

PC52046-1G PC52046-5G
77416-490EA 80.28 USD
77416-490 77416-492
Perfluorobutane-1-sulfonic anhydride 95%
Perfluorobutane-1-sulfonic anhydride
Formula: C₈F₁₈O₅S₂
MW: 582.184 g/mol
Boiling Pt: 84 °C
Density: 1.898 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD03427256
CAS Number: 36913-91-4
EINECS: 253-270-9
UN: 3264
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR