N-[1-(3-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

PC520401-100MG PC520401-1G PC520401-250MG
77416-478EA 21.26 USD
77416-478 77416-480 77416-482
N-[1-(3-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride, Technical Grade
N-[1-(3-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Formula: C₁₁H₁₂FN·ClH
MW: 213.679 g/mol
Storage Temperature: Freezer
MDL Number: MFCD31745723
CAS Number: 2379918-46-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR