Methyl chlorodifluoroacetate 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC5070-100G PC5070-25G PC5070-5G
77416-400EA 135.5 USD
77416-400 77416-402 77416-404
Methyl chlorodifluoroacetate 97%
Methyl chlorodifluoroacetate

Useful trifluoromethylating agent. Trifluoromethylation ofaryl and vinyl halides: Tetrahedron Letts.,32, 7689 (1991). Nitroaryl chlorides can also be trifluoromethylated.

Formula: C₃H₃ClF₂O₂
MW: 144.51
Boiling Pt: 79…81 °C
Density: 1.3701
Flash Pt: 19 °C
Storage Temperature: Ambient
MDL Number: MFCD00000775
CAS Number: 1514-87-0
EINECS: 216-154-9
UN: 2924
ADR: 3+8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR