4-Bromo-2-fluorophenyl isocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503015-1G PC503015-25G PC503015-5G
77416-330EA 54 USD
77416-330 77416-332 77416-334
4-Bromo-2-fluorophenyl isocyanate
4-Bromo-2-fluorophenyl isocyanate

  • May form small amount of sediment.

Formula: C₇H₃BrFNO
Storage Temperature: Refrigerator
CAS Number: 88112-75-8
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR