4-Bromo-2-(trifluoromethoxy)phenylisocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503013-1G PC503013-25G PC503013-5G
77416-320EA 21.26 USD
77416-320 77416-322 77416-324
4-Bromo-2-(trifluoromethoxy)phenylisocyanate
4-Bromo-2-(trifluoromethoxy)phenylisocyanate

May form small amount of sediment.

Formula: C₈H₃BrF₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01320693
CAS Number: 229957-06-6
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR