2-Bromo-5-(trifluoromethoxy)phenylisothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC502999-1G PC502999-25G PC502999-5G
77416-238EA 90.68 USD
77416-238 77416-240 77416-242
2-Bromo-5-(trifluoromethoxy)phenylisothiocyanate
2-Bromo-5-(trifluoromethoxy)phenylisothiocyanate
Formula: C₈H₃BrF₃NOS
MW: 298.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32200434
CAS Number: 2098276-18-5
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR