Methyl 4-(bromomethyl)-3-fluorobenzoate 95%

Supplier: APOLLO SCIENTIFIC

PC501461-1G PC501461-5G
77416-058EA 78.44 USD
77416-058 77416-060
Methyl 4-(bromomethyl)-3-fluorobenzoate 95%
Methyl 4-(bromomethyl)-3-fluorobenzoate
Formula: C₉H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD00273345
CAS Number: 128577-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR