α,α,α-Trifluoro-o-cresol 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC4830-100G PC4830-250G PC4830-25G
77415-738EA 58.02 USD
77415-738 77415-740 77415-742
α,α,α-Trifluoro-o-cresol 99%
α,α,α-Trifluoro-o-cresol
Formula: C₇H₅F₃O
Boiling Pt: 147…148 °C
Melting Pt: 45…46 °C
Density: 1.3 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00002222
CAS Number: 444-30-4
EINECS: 207-148-7
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR