α-Bromo-2-(trifluoromethoxy)toluene 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4829-100G PC4829-25G PC4829-5G
77415-728EA 191.54 USD
77415-728 77415-730 77415-732
α-Bromo-2-(trifluoromethoxy)toluene 97%
α-Bromo-2-(trifluoromethoxy)toluene
Formula: C₈H₆BrF₃O
Density: 1.598 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00236321
CAS Number: 198649-68-2
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR