1,1,2,3,3,3-Hexafluoropropyl ethyl ether 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 1,1,2,3,3,3-Hexafluoropropyl ether

PC4795-1G PC4795-25G PC4795-5G
77415-720EA 21.4 USD
77415-720 77415-722 77415-724
1,1,2,3,3,3-Hexafluoropropyl ethyl ether 97%
1,1,2,3,3,3-Hexafluoropropyl ethyl ether
Formula: C₅H₆F₆O
Boiling Pt: 64…71 °C
Density: 1.321 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236122
CAS Number: 380-34-7
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR