1-Fluoro-2-ethynylbenzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC5957-5G PC5957-1G PC5957-25G
77417-214EA 33.62 USD
77417-214 77417-210 77417-212
1-Fluoro-2-ethynylbenzene 97%
1-Fluoro-2-ethynylbenzene
Formula: C₈H₅F
Boiling Pt: 150 °C
Density: 1.06 g/cm³ (25 °C)
Flash Pt: 37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00799540
CAS Number: 766-49-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR