2-Fluoro-5-(trifluoromethoxy)aniline 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC5900-1G PC5900-25G PC5900-5G
77417-144EA 29.54 USD
77417-144 77417-146 77417-148
2-Fluoro-5-(trifluoromethoxy)aniline 99%
2-Fluoro-5-(trifluoromethoxy)aniline
Formula: C₇H₅F₄NO
MW: 195.12 g/mol
Boiling Pt: 64…65 °C/15 mmHg
Storage Temperature: Ambient
MDL Number: MFCD09832254
CAS Number: 116369-23-4
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR