Perfluorobutane-1-sulfonyl fluoride 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Perfluoro-1-butanesulphonyl fluoride, Nonafluorobutanesulphonyl fluoride

PC5887-250G PC5887-25G PC5887-100G
77417-134EA 87.62 USD
77417-134 77417-136 77417-132
Perfluorobutane-1-sulfonyl fluoride 95%
Perfluorobutane-1-sulphonyl fluoride
Formula: C₄F₁₀O₂S
MW: 302.09
Boiling Pt: 64…66 °C
Melting Pt: –110 °C
Density: 1.682 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007422
CAS Number: 375-72-4
EINECS: 206-792-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR