(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC5063L-1G PC5063L-250MG
77416-388EA 179.32 USD
77416-388 77416-390
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%
(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

For determination of enantiomeric excess of alcohols and amines by NMR.

Formula: C₁₀H₈ClF₃O₂
Boiling Pt: 213…214 °C
Density: 1.35 g/cm³ (25 °C)
Flash Pt: 89 °C
Storage Temperature: Freezer
MDL Number: MFCD00044400
CAS Number: 39637-99-5
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR