2-Nitro-4-(trifluoromethyl)anisole 97%

Supplier: APOLLO SCIENTIFIC

PC5060-100G PC5060-25G PC5060-5G
77416-378EA 135.5 USD
77416-378 77416-380 77416-382
2-Nitro-4-(trifluoromethyl)anisole 97%
2-Nitro-4-(trifluoromethyl)anisole
Formula: C₈H₆F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD00007099
CAS Number: 394-25-2
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR