3-Bromo-5-fluorophenylisothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503011-1G PC503011-25G PC503011-5G
77416-310EA 83.54 USD
77416-310 77416-312 77416-314
3-Bromo-5-fluorophenylisothiocyanate
3-Bromo-5-fluorophenylisothiocyanate
Formula: C₇H₃BrFNS
MW: 232.073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32200432
CAS Number: 1861560-52-2
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR