3-Bromo-4-fluorophenyl isothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503009-1G PC503009-25G PC503009-5G
77416-298EA 83.54 USD
77416-298 77416-300 77416-302
3-Bromo-4-fluorophenyl isothiocyanate
3-Bromo-4-fluorophenyl isothiocyanate
Formula: C₇H₃BrFNS
MW: 232.08 g/mol
Storage Temperature: Ambient
CAS Number: 710351-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR