2-Bromo-4-(trifluoromethoxy)phenylisothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC502996-1G PC502996-25G PC502996-5G
77416-220EA 83.54 USD
77416-220 77416-222 77416-224
2-Bromo-4-(trifluoromethoxy)phenylisothiocyanate
2-Bromo-4-(trifluoromethoxy)phenylisothiocyanate
Formula: C₈H₃BrF₃NOS
MW: 298.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800656
CAS Number: 948294-38-0
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR