2,2,2-Trifluoroethyl-2-fluoroacrylate

Supplier: APOLLO SCIENTIFIC
Warning

PC4975-1G PC4975-250MG
77415-972EA 48.9 USD
77415-972 77415-974
2,2,2-Trifluoroethyl-2-fluoroacrylate
2,2,2-Trifluoroethyl-2-fluoroacrylate
Formula: C₅H₄F₄O₂
Boiling Pt: 28 °C (30 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD04039290
CAS Number: 74359-10-7
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR