2-Fluoro-4-(methylsulfanyl)aniline

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Fluoro-4-(methylthio)aniline, 4-Amino-3-fluorothioanisole

PC49180-1G PC49180-25G PC49180-5G
77415-882EA 41.78 USD
77415-882 77415-884 77415-886
2-Fluoro-4-(methylsulfanyl)aniline
2-Fluoro-4-(methylsulfanyl)aniline
Formula: C₇H₈FNS
MW: 157.209 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18909665
CAS Number: 76180-33-1
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR