2-Bromo-5-(trifluoromethoxy)phenylacetylene 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Bromo-2-ethynyl-4-(trifluoromethoxy)benzene

PC48788-1G PC48788-250MG
77415-802EA 130.4 USD
77415-802 77415-804
2-Bromo-5-(trifluoromethoxy)phenylacetylene 95%
2-Bromo-5-(trifluoromethoxy)phenylacetylene
Formula: C₉H₄BrF₃O
MW: 265.027 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31726320
CAS Number: 2379918-32-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR