3,3,3-Trifluoropropionitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4553-1G PC4553-25G PC4553-5G
77415-622EA 63.16 USD
77415-622 77415-624 77415-626
3,3,3-Trifluoropropionitrile 97%
3,3,3-Trifluoropropionitrile
Formula: C₃H₂F₃N
Boiling Pt: 92 °C
Density: 1.279 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD07784277
CAS Number: 20530-38-5
UN: 3275
ADR: 6.1+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR