Ethyl 2,2,3,3-tetrafluoropropyl carbonate

Supplier: APOLLO SCIENTIFIC
Warning

PC450408-1G PC450408-5G
77415-546EA 96 USD
77415-546 77415-548
Ethyl 2,2,3,3-tetrafluoropropyl carbonate
Ethyl 2,2,3,3-tetrafluoropropyl carbonate
Formula: C₆H₈F₄O₃
MW: 204.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28334385
CAS Number: 277332-97-5
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR