2,2-Difluoroethyl 2,2,2-trifluoroethyl carbonate

Supplier: APOLLO SCIENTIFIC
Warning

PC450293-1G PC450293-5G
77415-538EA 185.92 USD
77415-538 77415-540
2,2-Difluoroethyl 2,2,2-trifluoroethyl carbonate
2,2-Difluoroethyl 2,2,2-trifluoroethyl carbonate
Formula: C₅H₅F₅O₃
MW: 208.083 g/mol
Boiling Pt: 77…78 °C (75 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD28334288
CAS Number: 1980085-24-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR