Tris(2,2-difluoroethyl)borate

Supplier: APOLLO SCIENTIFIC
Warning

PC450157-1G PC450157-250MG
77415-462EA 105.06 USD
77415-462 77415-464 77415-466
Tris(2,2-difluoroethyl)borate
Tris(2,2-difluoroethyl)borate
Formula: C₆H₉BF₆O₃
Storage Temperature: Ambient
MDL Number: MFCD19442172
CAS Number: 1396338-09-2
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR