4-Bromo-5-fluoro-2-iodobenzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC53385-1G PC53385-250MG PC53385-5G
77416-686EA 399.38 USD
77416-686 77416-688 77416-690
4-Bromo-5-fluoro-2-iodobenzonitrile 95%
4-Bromo-5-fluoro-2-iodobenzonitrile
Formula: C₇H₂BrFIN
MW: 325.904 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30297368
CAS Number: 2149601-45-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR