p-Tolylsulfur pentafluoride

Supplier: APOLLO SCIENTIFIC
Danger

PC49104-1G PC49104-250MG PC49104-5G
77415-848EA 265.76 USD
77415-848 77415-850 77415-852
p-Tolylsulfur pentafluoride
p-Tolylsulfur pentafluoride
Formula: C₇H₇F₅S
Boiling Pt: 80…81 °C/54 mmHg
Density: 1.414 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16817600
CAS Number: 203126-21-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR