4-(Difluoromethyl)benzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(Bromomethyl)benzal fluoride, 1-(Bromomethyl)-4-(difluoromethyl)benzene

PC48439-1G PC48439-250MG
77415-766EA 319.9 USD
77415-766 77415-768
4-(Difluoromethyl)benzyl bromide
4-(Difluoromethyl)benzyl bromide
Formula: C₈H₇BrF₂
Storage Temperature: Refrigerator
MDL Number: MFCD18791603
CAS Number: 873373-34-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR