α,α,α-Trifluoro-m-cresol 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC4840-100G PC4840-25G PC4840-500G
77415-756EA 23.34 USD
77415-756 77415-758 77415-760
α,α,α-Trifluoro-m-cresol 98%
α,α,α-Trifluoro-m-cresol
Formula: C₇H₅F₃O
Boiling Pt: 178 °C
Density: 1.333 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002299
CAS Number: 98-17-9
EINECS: 202-645-5
UN: 2927
ADR: 6.1+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR