(3,3,3-Trifluoropropyl)dimethoxymethylsilane 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Dimethoxy(methyl)(3,3,3-trifluoropropyl)silane, 1,1,1-Trifluoro-3-[dimethoxy(methyl)silyl]propane
PC4452-100G
PC4452-250G
PC4452-25G
77415-416EA
469.12
USD
77415-416
77415-418
77415-420
(3,3,3-Trifluoropropyl)dimethoxymethylsilane 97%
(3,3,3-Trifluoropropyl)dimethoxymethylsilane
Formula:
C₆H₁₃F₃O₂Si MW: 202.25 g/mol Boiling Pt: 85 °C/150 mmHg Density: 1.089 g/cm³ (25 °C) Flash Pt: 57.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053175 CAS Number: 358-67-8 EINECS: 206-619-4 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...