2-Fluoro-4-(trifluoromethyl)benzyl bromide 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC4376F-100G PC4376F-25G PC4376F-5G
77415-338EA 615.36 USD
77415-338 77415-340 77415-342
2-Fluoro-4-(trifluoromethyl)benzyl bromide 98%
2-Fluoro-4-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrF₄
Melting Pt: 28…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00061173
CAS Number: 239087-07-1
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR