2-Chloro-5-(trifluoromethyl)benzylamine 95%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC4327-1G
PC4327-25G
PC4327-5G
77415-260EA
21.28
USD
77415-260
77415-262
77415-264
2-Chloro-5-(trifluoromethyl)benzylamine 95%
2-Chloro-5-(trifluoromethyl)benzylamine
Formula:
C₈H₇ClF₃N MW: 209.6 g/mol Boiling Pt: 90…94 °C/13 mmHg Density: 1.33 g/cm³ (25 °C) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190123 CAS Number: 15996-78-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...