5-(Pentafluorothio)-1H-benzimidazole
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC49306-1G
PC49306-250MG
77415-906EA
263.32
USD
77415-906
77415-908
5-(Pentafluorothio)-1H-benzimidazole
5-(Pentafluorothio)-1H-benzimidazole
Formula:
C₇H₅F₅N₂S Storage Temperature: Ambient |
MDL Number:
MFCD22370117 CAS Number: 663179-60-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...