3-(Difluoromethyl)benzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Bromomethyl)-3-(difluoromethyl)benzene, 3-(Difluoromethyl)benzyl Bromide, α-Bromo-α',α'-difluoro-m-xylene, 3-(Bromomethyl)benzal fluoride

PC49200-1G PC49200-25G PC49200-5G
77415-894EA 101.88 USD
77415-894 77415-896 77415-898
3-(Difluoromethyl)benzyl bromide 97%
3-(Difluoromethyl)benzyl bromide
Formula: C₈H₇BrF₂
Storage Temperature: Refrigerator
MDL Number: MFCD18791600
CAS Number: 1263178-51-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR