3,3,3-Trifluoropropylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Amino-1,1,1-trifluoropropane

PC4603-1G PC4603-500MG PC4603-5G
77415-638EA 75.4 USD
77415-638 77415-640 77415-642
3,3,3-Trifluoropropylamine 97%
3,3,3-Trifluoropropylamine
Formula: C₃H₆F₃N
MW: 113.082 g/mol
Boiling Pt: 67…68 °C
Storage Temperature: Ambient
MDL Number: MFCD01694854
CAS Number: 460-39-9
UN: 2924
ADR: 3+8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR