2-Fluoro-4-(trifluoromethyl)benzylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (2-Fluoro-4-(trifluoromethyl)phenyl)methanamine

PC4374U-1G PC4374U-250MG PC4374U-500MG
77415-310EA 35.4 USD
77415-310 77415-312 77415-314
2-Fluoro-4-(trifluoromethyl)benzylamine 97%
2-Fluoro-4-(trifluoromethyl)benzylamine
Formula: C₈H₇F₄N
Storage Temperature: Ambient
MDL Number: MFCD00070807
CAS Number: 239087-05-9
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR