2-Fluoro-5-(trifluoromethyl)benzoyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4374O-100G PC4374O-25G PC4374O-5G
77415-300EA 339.26 USD
77415-300 77415-302 77415-304
2-Fluoro-5-(trifluoromethyl)benzoyl chloride 97%
2-Fluoro-5-(trifluoromethyl)benzoyl chloride
Formula: C₈H₃ClF₄O
Boiling Pt: 190…191 °C
Density: 1.499 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00061156
CAS Number: 207981-46-2
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR