3-Fluorostyrene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC4240-10G PC4240-1G PC4240-5G
77415-222EA 141.62 USD
77415-222 77415-224 77415-226
3-Fluorostyrene 98%
3-Fluorostyrene
Formula: C₈H₇F
MW: 122.14 g/mol
Boiling Pt: 31 °C/4 mmHg
Density: 1.025 g/cm³ (25 °C)
Flash Pt: 29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000341
CAS Number: 350-51-6
EINECS: 206-504-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR