4-Fluorobenzylboronic acid, pinacol ester 95+%

Supplier: APOLLO SCIENTIFIC

Synonyms: 2-(4-Fluorobenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC412402-1G PC412402-250MG
77415-152EA 195.62 USD
77415-152 77415-154
4-Fluorobenzylboronic acid, pinacol ester 95+%
4-Fluorobenzylboronic acid, pinacol ester
Formula: C₁₃H₁₈BFO₂
MW: 236.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10698519
CAS Number: 243145-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR