3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC3862-1G PC3862-5G
77414-984EA 64.18 USD
77414-984 77414-986
3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine 97%
3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine
Formula: C₆H₂BrF₄N
Density: 1.774 g/cm³ (25 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD08061304
CAS Number: 1031929-01-7
UN: 1992
ADR: 3+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR