2-Fluorophenylsulfur pentafluoride 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC49115-1G PC49115-250MG
77415-856EA 167.14 USD
77415-856 77415-858
2-Fluorophenylsulfur pentafluoride 97%
2-Fluorophenylsulphur pentafluoride
Formula: C₆H₄F₆S
Boiling Pt: 153…155 °C/637 mmHg
Storage Temperature: Ambient
MDL Number: MFCD18909664
CAS Number: 864230-02-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR