4-Nitro-3-(trifluoromethyl)phenol 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4860-1G PC4860-25G PC4860-5G
77415-776EA 21.26 USD
77415-776 77415-778 77415-780
4-Nitro-3-(trifluoromethyl)phenol 97%
4-Nitro-3-(trifluoromethyl)phenol
Formula: C₇H₄F₃NO₃
Boiling Pt: 135…138 °C/0.01 mmHg
Melting Pt: 72…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00014714
CAS Number: 88-30-2
EINECS: 201-818-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR